Anti-SH2D1A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: SH2D1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-430
- 77799-431
- Antigen Symbol:
- SH2D1A
- SH2D1A
- Antigen Name:
- SH2 domain-containing protein 1A protein (11-124AA)
- SH2 domain-containing protein 1A protein (11-124AA)
- Antigen Synonyms:
- DSHP|Duncan disease SH2 protein|Duncan disease SH2-protein|EBVS|IMD5|LYP|MTCP1|SAP|SAP/SH2D1A|SH2 domain containing 1A|SH2 domain protein 1A|SH2 domain-containing protein 1A|SH21A_HUMAN|SH2D1A|Signaling lymphocyte activation molecule associated protein|Signaling lymphocytic activation molecule-associated protein|SLAM associated protein|SLAM associated protein/SH2 domain protein 1A|SLAM-associated protein|T cell signal transduction molecule SAP|T-cell signal transduction molecule SAP|XLP|XLPD
- DSHP|Duncan disease SH2 protein|Duncan disease SH2-protein|EBVS|IMD5|LYP|MTCP1|SAP|SAP/SH2D1A|SH2 domain containing 1A|SH2 domain protein 1A|SH2 domain-containing protein 1A|SH21A_HUMAN|SH2D1A|Signaling lymphocyte activation molecule associated protein|Signaling lymphocytic activation molecule-associated protein|SLAM associated protein|SLAM associated protein/SH2 domain protein 1A|SLAM-associated protein|T cell signal transduction molecule SAP|T-cell signal transduction molecule SAP|XLP|XLPD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60880
- O60880
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together