Order Entry
United States
ContactUsLinkComponent
Anti-SSR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-SSR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SSR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77801-114
  • 77801-113
  • Antigen Symbol:
  • SSR2
  • SSR2
  • Antigen Name:
  • Translocon-associated protein subunit beta protein (18-149AA)
  • Translocon-associated protein subunit beta protein (18-149AA)
  • Antigen Synonyms:
  • SSR2|TRAPB|HSD25|Translocon-associated protein subunit beta|TRAP-beta|Signal sequence receptor subunit beta|SSR-beta
  • SSR2|TRAPB|HSD25|Translocon-associated protein subunit beta|TRAP-beta|Signal sequence receptor subunit beta|SSR-beta
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P43308
  • P43308
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together