Anti-TRAF1 Rabbit Polyclonal Antibody
Catalog # 77803-441
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TRAF1_HUMAN
- Antigen Symbol:TRAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13077
- Antigen Synonyms:4732496E14Rik|EBI6|EBV induced protein 6|Epstein-Barr virus-induced protein 6|MGC:10353|Tnf receptor associated factor 1|TNF receptor-associated factor 1|TRAF 1|TRAF1|TRAF1_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat