Anti-MRPL51 Rabbit Recombinant Polyclonal Antibody
Catalog # 77826-936
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:39S ribosomal protein L51, mitochondrial protein (32-128AA)
- Antigen Symbol:MRPL51
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q4U2R6
- Antigen Synonyms:39S ribosomal protein L51| 39S ribosomal protein L51 mitochondrial| bMRP-64| bMRP64| CDA09| HSPC241| L51mt| mitochondrial| mitochondrial ribosomal protein 64| mitochondrial ribosomal protein bMRP64| mitochondrial ribosomal protein L51| MRP-L51| MRP64| mrpl51| RM51_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: MRPL51
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human