Anti-NLRP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NLRP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-595
- 77836-596
- Antigen Symbol:
- NLRP2
- NLRP2
- Antigen Name:
- NACHT, LRR and PYD domains-containing protein 2 protein (1-220AA)
- NACHT, LRR and PYD domains-containing protein 2 protein (1-220AA)
- Antigen Synonyms:
- CLR19.9|FLJ20510|Leucine rich repeat and PYD containing protein 2|LRR and PYD domains-containing protein 2|NACHT|NACHT domain|NACHT leucine rich repeat and PYD containing 2|NACHT LRR and PYD domains containing protein 2|NALP2|NALP2_HUMAN|NBS1|NLR family pyrin domain containing 2||Nucleotide binding oligomerization domain leucine rich repeat and pyrin domain containing 2|Nucleotide-binding site protein 1|PAN1|PYPAF2|PYRIN containing APAF1 like protein2|PYRIN domain and NACHT domain-containing protein 1|PYRIN-containing APAF1-like protein 2
- CLR19.9|FLJ20510|Leucine rich repeat and PYD containing protein 2|LRR and PYD domains-containing protein 2|NACHT|NACHT domain|NACHT leucine rich repeat and PYD containing 2|NACHT LRR and PYD domains containing protein 2|NALP2|NALP2_HUMAN|NBS1|NLR family pyrin domain containing 2||Nucleotide binding oligomerization domain leucine rich repeat and pyrin domain containing 2|Nucleotide-binding site protein 1|PAN1|PYPAF2|PYRIN containing APAF1 like protein2|PYRIN domain and NACHT domain-containing protein 1|PYRIN-containing APAF1-like protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NX02
- Q9NX02
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together