Anti-FOXF1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: FOXF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77825-937
- 77825-936
- Antigen Symbol:
- FOXF1
- FOXF1
- Antigen Name:
- Synthetic peptide of FOXF1
- Synthetic peptide of FOXF1
- Antigen Synonyms:
- AI450827|FKH L5|FKHL 5|FKHL5|Forkhead (Drosophila) like 5|Forkhead box F1|Forkhead box protein F1|Forkhead drosophila homolog like 5|Forkhead like 5|Forkhead related activator 1|Forkhead related protein FKHL5|Forkhead related transcription factor 1|Forkhead-related activator 1|Forkhead-related protein FKHL5|Forkhead-related transcription factor 1|FOX F1|FOXF 1|foxf1|FOXF1_HUMAN|FREAC 1|FREAC-1|FREAC1|HFH 8|MGC105125
- AI450827|FKH L5|FKHL 5|FKHL5|Forkhead (Drosophila) like 5|Forkhead box F1|Forkhead box protein F1|Forkhead drosophila homolog like 5|Forkhead like 5|Forkhead related activator 1|Forkhead related protein FKHL5|Forkhead related transcription factor 1|Forkhead-related activator 1|Forkhead-related protein FKHL5|Forkhead-related transcription factor 1|FOX F1|FOXF 1|foxf1|FOXF1_HUMAN|FREAC 1|FREAC-1|FREAC1|HFH 8|MGC105125
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12946
- Q12946
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together