Order Entry
United States
ContactUsLinkComponent
Anti-TANK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-TANK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TANK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77801-843
  • 77801-842
  • Antigen Symbol:
  • TANK
  • TANK
  • Antigen Name:
  • TRAF family member-associated NF-kappa-B activator protein (4-98AA)
  • TRAF family member-associated NF-kappa-B activator protein (4-98AA)
  • Antigen Synonyms:
  • I TRAF|I-TRAF|ITRAF|Tank|TANK_HUMAN|TRAF family member associated NF KAPPA B activator|TRAF family member associated NFKB activator|TRAF family member-associated NF-kappa-B activator|TRAF interacting protein|TRAF interacting protein TANK isoform a|TRAF interacting protein TANK isoform b|TRAF-interacting protein|TRAF2
  • I TRAF|I-TRAF|ITRAF|Tank|TANK_HUMAN|TRAF family member associated NF KAPPA B activator|TRAF family member associated NFKB activator|TRAF family member-associated NF-kappa-B activator|TRAF interacting protein|TRAF interacting protein TANK isoform a|TRAF interacting protein TANK isoform b|TRAF-interacting protein|TRAF2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q92844
  • Q92844
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together