Anti-IGFBP7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: IGFBP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77812-801
- 77812-802
- Antigen Symbol:
- IGFBP7
- IGFBP7
- Antigen Name:
- Synthetic peptide of IGFBP7
- Synthetic peptide of IGFBP7
- Antigen Synonyms:
- AGM|Angiomodulin|FSTL2|IBP-7|IBP7|IBP7_HUMAN|IGF binding protein 7|IGF-binding protein 7|IGFBP rP1|IGFBP-7|IGFBP-rP1|IGFBP7|IGFBPRP1|Insulin like growth factor binding protein 7|Insulin-like growth factor-binding protein 7|MAC25|MAC25 protein|PGI2 stimulating factor|PGI2-stimulating factor|Prostacyclin stimulating factor|Prostacyclin-stimulating factor|PSF|RAMSVPS|TAF|Tumor-derived adhesion factor
- AGM|Angiomodulin|FSTL2|IBP-7|IBP7|IBP7_HUMAN|IGF binding protein 7|IGF-binding protein 7|IGFBP rP1|IGFBP-7|IGFBP-rP1|IGFBP7|IGFBPRP1|Insulin like growth factor binding protein 7|Insulin-like growth factor-binding protein 7|MAC25|MAC25 protein|PGI2 stimulating factor|PGI2-stimulating factor|Prostacyclin stimulating factor|Prostacyclin-stimulating factor|PSF|RAMSVPS|TAF|Tumor-derived adhesion factor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16270
- Q16270
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together