Anti-CD4L-2 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: CD4L-2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Grass carp
- Catalog No:
- 77814-344
- 77814-343
- Antigen Symbol:
- CD4L-2
- CD4L-2
- Antigen Name:
- Grass carp CD4-like protein 2 protein (59-77AA)
- Grass carp CD4-like protein 2 protein (59-77AA)
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Grass carp
- Grass carp
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A0A1B2FIB0
- A0A1B2FIB0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together