Anti-UNG Rabbit Polyclonal Antibody
Catalog # 77804-023
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Uracil-DNA glycosylase
- Antigen Symbol:UNG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P13051
- Antigen Synonyms:DGU|DKFZp781L1143|HIGM 4|HIGM4|OTTHUMP00000240527|OTTHUMP00000240528|OTTHUMP00000240529|UDG|UNG 1|UNG 15|ung|UNG_HUMAN|UNG1|UNG15|UNG2|Uracil DNA glycosylase 1|Uracil DNA glycosylase 2|Uracil DNA glycosylase|Uracil-DNA glycosylase
- Storage Buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat