Anti-RALA Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: RALA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-596
- 77796-595
- Antigen Symbol:
- RALA
- RALA
- Antigen Name:
- Ras-related protein Ral-A protein (1-203AA)
- Ras-related protein Ral-A protein (1-203AA)
- Antigen Synonyms:
- MGC48949|Ral a|Ral A protein|RAL|RALA|RALA_HUMAN|Ras family small GTP binding protein RALA|RAS like protein A|Ras related protein RalA|Ras-related protein Ral-A|v ral simian leukemia viral oncogene homolog A (ras related)|v ral simian leukemia viral oncogene homolog A
- MGC48949|Ral a|Ral A protein|RAL|RALA|RALA_HUMAN|Ras family small GTP binding protein RALA|RAS like protein A|Ras related protein RalA|Ras-related protein Ral-A|v ral simian leukemia viral oncogene homolog A (ras related)|v ral simian leukemia viral oncogene homolog A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11233
- P11233
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together