Anti-TWIST1 Rabbit Polyclonal Antibody
Catalog # 77803-737
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TWST1_HUMAN
- Antigen Symbol:TWIST1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q15672
- Antigen Synonyms:ACS3|B-HLH DNA binding protein|bHLHa38|BPES2|BPES3|Class A basic helix-loop-helix protein 38|CRS|CRS1|CSO|H-twist|OTTHUMP00000116043|SCS|Twist basic helix loop helix transcription factor 1|Twist family bHLH transcription factor 1|Twist homolog 1 (Drosophila)|Twist homolog 1|TWIST homolog of drosophila|Twist related protein 1|Twist-related protein 1|TWIST1|TWST1_HUMAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TWIST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat