Anti-MEST Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MEST
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-553
- 77827-554
- Antigen Symbol:
- MEST
- MEST
- Antigen Name:
- Mesoderm-specific transcript homolog protein (146-335AA)
- Mesoderm-specific transcript homolog protein (146-335AA)
- Antigen Synonyms:
- DKFZp686L18234|EC 3.-.-.-|Mesoderm specific transcript (mouse) homolog|Mesoderm specific transcript homolog (mouse)|Mesoderm-specific transcript homolog protein|MEST|MEST_HUMAN|MGC111102|MGC8703|OTTHUMP00000210362|OTTHUMP00000210364|OTTHUMP00000210367|Paternally expressed gene 1|Paternally-expressed gene 1 protein|PEG1
- DKFZp686L18234|EC 3.-.-.-|Mesoderm specific transcript (mouse) homolog|Mesoderm specific transcript homolog (mouse)|Mesoderm-specific transcript homolog protein|MEST|MEST_HUMAN|MGC111102|MGC8703|OTTHUMP00000210362|OTTHUMP00000210364|OTTHUMP00000210367|Paternally expressed gene 1|Paternally-expressed gene 1 protein|PEG1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5EB52
- Q5EB52
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together