Anti-PITRM1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77827-133
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Presequence protease, mitochondrial protein (758-1037AA)
- Antigen Symbol:PITRM1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q5JRX3
- Antigen Synonyms:hMP1|hPreP|KIAA1104|Metalloprotease 1|MGC138192|MGC141929|mitochondrial|MP1|Pitrilysin metallopeptidase 1|Pitrilysin metalloproteinase 1|PITRM 1|PITRM1|PreP|PreP peptidasome|PREP_HUMAN|Presequence protease|Presequence protease mitochondrial
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PITRM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse