Anti-NDFIP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDFIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-858
- 77842-857
- Antigen Symbol:
- NDFIP1
- NDFIP1
- Antigen Name:
- NEDD4 family-interacting protein 1 protein (2-116AA)
- NEDD4 family-interacting protein 1 protein (2-116AA)
- Antigen Synonyms:
- 0610010M22Rik|Breast cancer associated protein SGA 1M|Breast cancer-associated protein SGA-1M|MGC10924|N4WBP5|Ndfip1|Nedd4 family interacting protein 1|NEDD4 family-interacting protein 1|NEDD4 WW domain binding protein 5|NEDD4 WW domain-binding protein 5|NFIP1_HUMAN|Putative MAPK activating protein PM13|Putative MAPK-activating protein PM13|Putative NF kappa B activating protein 164|Putative NF-kappa-B-activating protein 164|Putative NFKB and MAPK activating protein|Putative NFKB and MAPK-activating protein
- 0610010M22Rik|Breast cancer associated protein SGA 1M|Breast cancer-associated protein SGA-1M|MGC10924|N4WBP5|Ndfip1|Nedd4 family interacting protein 1|NEDD4 family-interacting protein 1|NEDD4 WW domain binding protein 5|NEDD4 WW domain-binding protein 5|NFIP1_HUMAN|Putative MAPK activating protein PM13|Putative MAPK-activating protein PM13|Putative NF kappa B activating protein 164|Putative NF-kappa-B-activating protein 164|Putative NFKB and MAPK activating protein|Putative NFKB and MAPK-activating protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BT67
- Q9BT67
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together