Anti-rbtD Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: rbtD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterobacter aerogenes
Specifications
- Catalog No:
- 77817-352
- 77817-353
- Antigen Symbol:
- rbtD
- rbtD
- Antigen Name:
- Recombinant Enterobacter aerogenes Ribitol 2-dehydrogenase protein (1-249AA)
- Recombinant Enterobacter aerogenes Ribitol 2-dehydrogenase protein (1-249AA)
- Antigen Synonyms:
- rbtD|Ribitol 2-dehydrogenase|RDH|EC 1.1.1.56
- rbtD|Ribitol 2-dehydrogenase|RDH|EC 1.1.1.56
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Enterobacter aerogenes
- Enterobacter aerogenes
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P00335
- P00335
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together