Anti-SULT1B1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SULT1B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-556
- 77801-555
- Antigen Symbol:
- SULT1B1
- SULT1B1
- Antigen Name:
- Sulfotransferase family cytosolic 1B member 1 protein (1-271AA)
- Sulfotransferase family cytosolic 1B member 1 protein (1-271AA)
- Antigen Synonyms:
- Dopa/tyrosine sulfotransferase|MGC13356|ST1B1|ST1B1_HUMAN|ST1B2|Sulfotransferase 1B1|Sulfotransferase 1B2|Sulfotransferase family 1B member 1|Sulfotransferase family cytosolic 1B member 1|SULT1B1|SULT1B2|Thyroid hormone sulfotransferase
- Dopa/tyrosine sulfotransferase|MGC13356|ST1B1|ST1B1_HUMAN|ST1B2|Sulfotransferase 1B1|Sulfotransferase 1B2|Sulfotransferase family 1B member 1|Sulfotransferase family cytosolic 1B member 1|SULT1B1|SULT1B2|Thyroid hormone sulfotransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43704
- O43704
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together