Anti-RASGRF1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RASGRF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-526
- 77823-525
- Antigen Symbol:
- RASGRF1
- RASGRF1
- Antigen Name:
- Ras-specific guanine nucleotide-releasing factor 1 protein (1-190AA)
- Ras-specific guanine nucleotide-releasing factor 1 protein (1-190AA)
- Antigen Synonyms:
- CDC25| CDC25L| GNRP| GRF1| GRF55| Guanine nucleotide exchange factor| Guanine nucleotide releasing factor 55 kD| Guanine nucleotide releasing protein| guanine nucleotide-releasing factor 1| Guanine nucleotide-releasing protein| H GRF55| HGRF55| PP13187| Ras protein specific guanine nucleotide releasing factor 1| Ras specific guanine nucleotide releasing factor| Ras specific guanine nucleotide releasing factor CDC25 homolog| Ras specific nucleotide exchange factor CDC25| Ras-GRF1| Ras-specific guanine nucleotide-releasing factor 1| Ras-specific nucleotide exchange factor CDC25| RASGRF1| RGRF1_HUMAN
- CDC25| CDC25L| GNRP| GRF1| GRF55| Guanine nucleotide exchange factor| Guanine nucleotide releasing factor 55 kD| Guanine nucleotide releasing protein| guanine nucleotide-releasing factor 1| Guanine nucleotide-releasing protein| H GRF55| HGRF55| PP13187| Ras protein specific guanine nucleotide releasing factor 1| Ras specific guanine nucleotide releasing factor| Ras specific guanine nucleotide releasing factor CDC25 homolog| Ras specific nucleotide exchange factor CDC25| Ras-GRF1| Ras-specific guanine nucleotide-releasing factor 1| Ras-specific nucleotide exchange factor CDC25| RASGRF1| RGRF1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13972
- Q13972
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together