Anti-TECPR2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: TECPR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-197
- 77802-196
- Antigen Symbol:
- TECPR2
- TECPR2
- Antigen Name:
- Tectonin beta-propeller repeat-containing protein 2 protein (516-783AA)
- Tectonin beta-propeller repeat-containing protein 2 protein (516-783AA)
- Antigen Synonyms:
- TECPR2|KIAA0297|KIAA0329|Tectonin beta-propeller repeat-containing protein 2|WD repeat-containing protein KIAA0329/KIAA0297
- TECPR2|KIAA0297|KIAA0329|Tectonin beta-propeller repeat-containing protein 2|WD repeat-containing protein KIAA0329/KIAA0297
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15040
- O15040
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together