Anti-AFDN Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: AFDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77814-631
- 77814-630
- Antigen Symbol:
- AFDN
- AFDN
- Antigen Name:
- Synthetic peptide of MLLT4
- Synthetic peptide of MLLT4
- Antigen Synonyms:
- AF6|AFAD_HUMAN|Afadin|ALL1-fused gene from chromosome 6 protein|Mllt4|Protein Af-6|Protein AF6
- AF6|AFAD_HUMAN|Afadin|ALL1-fused gene from chromosome 6 protein|Mllt4|Protein Af-6|Protein AF6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55196
- P55196
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together