Anti-TRIM14 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TRIM14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77820-780
- 77820-779
- Antigen Symbol:
- TRIM14
- TRIM14
- Antigen Name:
- Synthetic peptide of TRIM14
- Synthetic peptide of TRIM14
- Antigen Synonyms:
- 5830400N10Rik|KIAA0129|pub|TRI14_HUMAN|TRIM14|tripartite motif protein 14|tripartite motif protein TRIM14|tripartite motif-containing 14|Tripartite motif-containing protein 14
- 5830400N10Rik|KIAA0129|pub|TRI14_HUMAN|TRIM14|tripartite motif protein 14|tripartite motif protein TRIM14|tripartite motif-containing 14|Tripartite motif-containing protein 14
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14142
- Q14142
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together