Anti-NDUFA2 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NDUFA2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-120
- 77806-119
- Antigen Symbol:
- NDUFA2
- NDUFA2
- Antigen Name:
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2 protein (4-99AA)
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2 protein (4-99AA)
- Antigen Synonyms:
- B8|CI B8|CI-B8|Complex I B8|Complex I-B8|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 2 8kDa|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 2|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2|NADH ubiquinone oxidoreductase B8 subunit|NADH-ubiquinone oxidoreductase B8 subunit|NDUA2_HUMAN|NDUFA 2|Ndufa2
- B8|CI B8|CI-B8|Complex I B8|Complex I-B8|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 2 8kDa|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 2|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2|NADH ubiquinone oxidoreductase B8 subunit|NADH-ubiquinone oxidoreductase B8 subunit|NDUA2_HUMAN|NDUFA 2|Ndufa2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43678
- O43678
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together