Anti-QRSL1 Rabbit Polyclonal Antibody
Catalog # 77796-295
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Glutaminyl-tRNA synthase-like protein 1
- Antigen Symbol:QRSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H0R6
- Antigen Synonyms:QRSL1| Glutamyl-tRNA(Gln) amidotransferase subunit A| mitochondrial| Glu-AdT subunit A| EC 6.3.5.7| Glutaminyl-tRNA synthase-like protein 1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: QRSL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat