Anti-SFPQ Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SFPQ
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-306
- 77799-307
- Antigen Symbol:
- SFPQ
- SFPQ
- Antigen Name:
- Splicing factor, proline- and glutamine-rich protein (499-598AA)
- Splicing factor, proline- and glutamine-rich protein (499-598AA)
- Antigen Synonyms:
- 100 kDa DNA pairing protein|100 kDa DNA-pairing protein|100 kDa subunit|DNA binding p52/p100 complex 100 kDa subunit|DNA-binding p52/p100 complex|hPOMp100|Polypyrimidine tract binding protein associated splicing factor|Polypyrimidine tract-binding protein-associated-splicing factor|POMP100|PPP1R140|proline- and glutamine-rich|Protein phosphatase 1 regulatory subunit 140|PSF|PTB associated splicing factor|PTB-associated-splicing factor|Sfpq|SFPQ_HUMAN|Splicing factor|Splicing factor proline and glutamine rich|Splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)|Splicing factor proline/glutamine rich
- 100 kDa DNA pairing protein|100 kDa DNA-pairing protein|100 kDa subunit|DNA binding p52/p100 complex 100 kDa subunit|DNA-binding p52/p100 complex|hPOMp100|Polypyrimidine tract binding protein associated splicing factor|Polypyrimidine tract-binding protein-associated-splicing factor|POMP100|PPP1R140|proline- and glutamine-rich|Protein phosphatase 1 regulatory subunit 140|PSF|PTB associated splicing factor|PTB-associated-splicing factor|Sfpq|SFPQ_HUMAN|Splicing factor|Splicing factor proline and glutamine rich|Splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)|Splicing factor proline/glutamine rich
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P23246
- P23246
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together