Anti-NDUFB11 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFB11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-569
- 77837-568
- Antigen Symbol:
- NDUFB11
- NDUFB11
- Antigen Name:
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial protein (30-88AA)
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial protein (30-88AA)
- Antigen Synonyms:
- NDUFB11| UNQ111/PRO1064NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11| mitochondrial| Complex I-ESSS| CI-ESSS| NADH-ubiquinone oxidoreductase ESSS subunit| Neuronal protein 17.3| Np17.3| p17.3
- NDUFB11| UNQ111/PRO1064NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11| mitochondrial| Complex I-ESSS| CI-ESSS| NADH-ubiquinone oxidoreductase ESSS subunit| Neuronal protein 17.3| Np17.3| p17.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NX14
- Q9NX14
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together