Order Entry
United States
ContactUsLinkComponent
Anti-EIF5A2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-EIF5A2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-EIF5A2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77838-232
  • 77838-231
  • Antigen Symbol:
  • EIF5A2
  • EIF5A2
  • Antigen Name:
  • Eukaryotic translation initiation factor 5A-2 protein (2-153AA)
  • Eukaryotic translation initiation factor 5A-2 protein (2-153AA)
  • Antigen Synonyms:
  • 9630038B20|eIF 5A 2|eIF 5A2|eIF-4D|eIF-5A-2|eIF-5A2|EIF5A 2|EIF5A2|eIF5AII|Eukaryotic initiation factor 5A isoform 2|Eukaryotic translation initiation factor 5A-2|Eukaryotic translation initiation factor 5A2|IF5A2_HUMAN
  • 9630038B20|eIF 5A 2|eIF 5A2|eIF-4D|eIF-5A-2|eIF-5A2|EIF5A 2|EIF5A2|eIF5AII|Eukaryotic initiation factor 5A isoform 2|Eukaryotic translation initiation factor 5A-2|Eukaryotic translation initiation factor 5A2|IF5A2_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9GZV4
  • Q9GZV4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together