To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Rab3 GTPase-activating protein catalytic subunit protein (527-653AA)
- Antigen Symbol:RAB3GAP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q15042
- Antigen Synonyms:DKFZp434A012|KIAA0066|P130|Rab3 GAP|Rab3 GAP p130|RAB3 GTPase activating protein 130 kDa subunit|Rab3 GTPase activating protein catalytic subunit|RAB3 GTPase activating protein subunit 1 (catalytic)|RAB3 GTPase-activating protein 130 kDa subunit|Rab3 GTPase-activating protein catalytic subunit|Rab3-GAP|Rab3-GAP p130|RAB3GAP|rab3gap1|RAB3GAP130|RB3GP_HUMAN|WARBM1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: RAB3GAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human