To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:MOB kinase activator 1B protein (2-216AA)
- Antigen Symbol:MOB1B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q7L9L4
- Antigen Synonyms:MATS 2|MATS2|MGC33910|Mob 1A|Mob 1B|MOB 4A|MOB kinase activator 1B|Mob1 homolog 1A|MOB1 Mps One Binder homolog B|MOB1 Mps one binder kinase activator like 1A|Mob1A|Mob1B|MOBKL 1A|MOBKL1A|MOL1A_HUMAN|Mps one binder kinase activator like 1A|Mps one binder kinase activator-like 1A|Protein Mob4A
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:500 to 1:2000, IF: 1:200 to 1:500, IP: 1:200 to 1:2000
Type:
Antigen: MOB1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human