Anti-SOCS1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:10000, WB:1:500 to 1:2000, IHC:1:100 to 1:300
Type:
Antigen: SOCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77813-515
- 77813-514
- Antigen Symbol:
- SOCS1
- SOCS1
- Antigen Name:
- Synthetic peptide of SOCS1
- Synthetic peptide of SOCS1
- Antigen Synonyms:
- CISH 1|CISH1|Cytokine inducible SH2 protein 1|JAB|JAK binding protein|JAK-binding protein|Janus kinase binding protein|SOCS 1|SOCS-1|Socs1|SOCS1_HUMAN|SSI 1|SSI-1|SSI1|STAT induced STAT inhibitor 1|STAT-induced STAT inhibitor 1|Suppressor of cytokine signaling 1|Supressor of cytokine signalling 1|TEC interacting protein 3|Tec-interacting protein 3|TIP 3|TIP-3|TIP3
- CISH 1|CISH1|Cytokine inducible SH2 protein 1|JAB|JAK binding protein|JAK-binding protein|Janus kinase binding protein|SOCS 1|SOCS-1|Socs1|SOCS1_HUMAN|SSI 1|SSI-1|SSI1|STAT induced STAT inhibitor 1|STAT-induced STAT inhibitor 1|Suppressor of cytokine signaling 1|Supressor of cytokine signalling 1|TEC interacting protein 3|Tec-interacting protein 3|TIP 3|TIP-3|TIP3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15524
- O15524
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together