Anti-NLRC5 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NLRC5
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-234
- 77832-235
- Antigen Symbol:
- NLRC5
- NLRC5
- Antigen Name:
- Protein NLRC5 protein (95-208AA)
- Protein NLRC5 protein (95-208AA)
- Antigen Synonyms:
- Caterpiller protein 16.1|CLR16.1|FLJ21709|FLJ39711|NLR family, CARD domain containing 5|NLR family, CARD-containing 5|NLR family, caspase recruitment domain-containing 5|NLRC5|NLRC5_HUMAN|NOD-like receptor C5|NOD27|NOD4|Nucleotide-binding oligomerization domain protein 27|Nucleotide-binding oligomerization domain protein 4|nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5|nucleotide-binding oligomerization domains 27|OTTHUMP00000164675|Protein Caterpiller 16.1|Protein NLRC5
- Caterpiller protein 16.1|CLR16.1|FLJ21709|FLJ39711|NLR family, CARD domain containing 5|NLR family, CARD-containing 5|NLR family, caspase recruitment domain-containing 5|NLRC5|NLRC5_HUMAN|NOD-like receptor C5|NOD27|NOD4|Nucleotide-binding oligomerization domain protein 27|Nucleotide-binding oligomerization domain protein 4|nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5|nucleotide-binding oligomerization domains 27|OTTHUMP00000164675|Protein Caterpiller 16.1|Protein NLRC5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WI3
- Q86WI3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together