Anti-HSPE1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: HSPE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Catalog No:
- 77820-105
- 77820-104
- Antigen Symbol:
- HSPE1
- HSPE1
- Antigen Name:
- Synthetic peptide of HSPE1
- Synthetic peptide of HSPE1
- Antigen Synonyms:
- 10 kDa chaperonin|10 kDa heat shock protein mitochondrial|10 kDa heat shock protein|mitochondrial|CH10_HUMAN|Chaperonin 10|Chaperonin 10 homolog|CPN10|cpn10 homolog|Early pregnancy factor|Early-pregnancy factor|EPF|GROES|GroES homolog|Heat shock 10kD protein 1 chaperonin 10|Heat shock 10kDa protein 1|Heat shock 10kDa protein 1 chaperonin 10|Heat shock protein family E (Hsp10) member|Heat-shock 10-kD protein|Hsp10|Hspe1
- 10 kDa chaperonin|10 kDa heat shock protein mitochondrial|10 kDa heat shock protein|mitochondrial|CH10_HUMAN|Chaperonin 10|Chaperonin 10 homolog|CPN10|cpn10 homolog|Early pregnancy factor|Early-pregnancy factor|EPF|GROES|GroES homolog|Heat shock 10kD protein 1 chaperonin 10|Heat shock 10kDa protein 1|Heat shock 10kDa protein 1 chaperonin 10|Heat shock protein family E (Hsp10) member|Heat-shock 10-kD protein|Hsp10|Hspe1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61604
- P61604
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together