Order Entry
United States
ContactUsLinkComponent
 
Anti-DET1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
 
undefined
Anti-DET1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product

 

  • Catalog No:
  • 77831-309
  • 77831-310
  • Antigen Symbol:
  • DET1
  • DET1
  • Antigen Name:
  • DET1 homolog protein (5-190AA)
  • DET1 homolog protein (5-190AA)
  • Antigen Synonyms:
  • DET1|DET1 homolog|De-etiolated-1 homolog
  • DET1|DET1 homolog|De-etiolated-1 homolog
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q7L5Y6
  • Q7L5Y6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together