Anti-TMLHE Rabbit Polyclonal Antibody
77811-164
New Product
- Antigen Name:XAP130
- Antigen Symbol:TMLHE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NVH6
- Antigen Synonyms:AUTSX6|BBOX2|Butyrobetaine (gamma) 2 oxoglutarate dioxygenase (gamma butyrobetaine hydroxylase) 2|Epsilon-trimethyllysine 2-oxoglutarate dioxygenase|Epsilon-trimethyllysine hydroxylase|FLJ10727|TML dioxygenase|TML hydroxylase|TML-alpha-ketoglutarate dioxygenase|TMLD|TMLH|TMLH_HUMAN|TMLHE|TMLHED|Trimethyllysine dioxygenase, mitochondrial|Trimethyllysine hydroxylase epsilon|XAP130
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TMLHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat