Anti-POU3F4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: POU3F4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-899
- 77794-900
- Antigen Symbol:
- POU3F4
- POU3F4
- Antigen Name:
- POU domain, class 3, transcription factor 4 protein (22-134AA)
- POU domain, class 3, transcription factor 4 protein (22-134AA)
- Antigen Synonyms:
- Brain specific homeobox POU domain protein 4|Brain-4|Brain-specific homeobox/POU domain protein 4|BRAIN4|Brn-4|BRN4|class 3|DFN3|DFNX2|Oct-9|Octamer-binding protein 9|Octamer-binding transcription factor 9|OTF-9|OTF9|PO3F4_HUMAN|POU class 3 homeobox 4|POU domain|POU domain class 3 transcription factor 4|Pou3f4|transcription factor 4
- Brain specific homeobox POU domain protein 4|Brain-4|Brain-specific homeobox/POU domain protein 4|BRAIN4|Brn-4|BRN4|class 3|DFN3|DFNX2|Oct-9|Octamer-binding protein 9|Octamer-binding transcription factor 9|OTF-9|OTF9|PO3F4_HUMAN|POU class 3 homeobox 4|POU domain|POU domain class 3 transcription factor 4|Pou3f4|transcription factor 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49335
- P49335
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together