Anti-ftsZ Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ftsZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Wolbachia pipientis
Specifications
- Catalog No:
- 77816-434
- 77816-433
- Antigen Symbol:
- ftsZ
- ftsZ
- Antigen Name:
- Recombinant Wolbachia pipientis Cell division protein FtsZ protein (1-315AA)
- Recombinant Wolbachia pipientis Cell division protein FtsZ protein (1-315AA)
- Antigen Synonyms:
- ftsZCell division protein FtsZ|Fragment
- ftsZCell division protein FtsZ|Fragment
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Wolbachia pipientis
- Wolbachia pipientis
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50907
- P50907
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together