Anti-MAT2B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MAT2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-173
- 77841-172
- Antigen Symbol:
- MAT2B
- MAT2B
- Antigen Name:
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Antigen Synonyms:
- 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
- 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZL9
- Q9NZL9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together