Anti-GSTO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:100 to 1:300
Type:
Antigen: GSTO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-199
- 77830-198
- Antigen Symbol:
- GSTO1
- GSTO1
- Antigen Name:
- Fusion protein of GSTO1
- Fusion protein of GSTO1
- Antigen Synonyms:
- AA407097|AI194287|AU018802|DKFZp686H13163|EC 2.5.1.18|Glutathione S transferase omega 1|Glutathione S-transferase omega 1-1|Glutathione S-transferase omega|Glutathione S-transferase omega-1|Glutathione transferase omega 1|Glutathione-dependent dehydroascorbate reductase|Glutathione-S-transferase like|Glutathione-S-transferase omega 1|GSTO 1-1|GSTO-1|GSTO1 1|Gsto1|GSTO1_HUMAN|GSTTLp28|GSTX|Gtsttl|MGC94845|MMA(V) reductase|Monomethylarsonic acid reductase|OTTHUMP00000020429|p28|S-(Phenacyl)glutathione reductase|SPG-R
- AA407097|AI194287|AU018802|DKFZp686H13163|EC 2.5.1.18|Glutathione S transferase omega 1|Glutathione S-transferase omega 1-1|Glutathione S-transferase omega|Glutathione S-transferase omega-1|Glutathione transferase omega 1|Glutathione-dependent dehydroascorbate reductase|Glutathione-S-transferase like|Glutathione-S-transferase omega 1|GSTO 1-1|GSTO-1|GSTO1 1|Gsto1|GSTO1_HUMAN|GSTTLp28|GSTX|Gtsttl|MGC94845|MMA(V) reductase|Monomethylarsonic acid reductase|OTTHUMP00000020429|p28|S-(Phenacyl)glutathione reductase|SPG-R
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78417
- P78417
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together