Anti-KLHL26 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KLHL26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-512
- 77827-511
- Antigen Symbol:
- KLHL26
- KLHL26
- Antigen Name:
- Kelch-like protein 26 protein (366-615AA)
- Kelch-like protein 26 protein (366-615AA)
- Antigen Synonyms:
- C630013N10Rik|FLJ11078|Kelch like 26 (Drosophila)|Kelch like protein 26|Kelch-like protein 26|KLH26_HUMAN|Klkl26
- C630013N10Rik|FLJ11078|Kelch like 26 (Drosophila)|Kelch like protein 26|Kelch-like protein 26|KLH26_HUMAN|Klkl26
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53HC5
- Q53HC5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together