Anti-SERINC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SERINC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-089
- 77799-090
- Antigen Symbol:
- SERINC1
- SERINC1
- Antigen Name:
- Serine incorporator 1 protein (331-387AA)
- Serine incorporator 1 protein (331-387AA)
- Antigen Synonyms:
- 1500011D18Rik|AI315070|AIGP 2|KIAA1253|LRRGT00191|mKIAA1253|SERC1_HUMAN|SERINC1|Serine incorporator 1|TDE1L|TDE2|TMS 2|Tms2|Tumor differentially expressed 1 protein like|Tumor differentially expressed protein 1-like|Tumor differentially expressed protein 2|UNQ396/PRO732
- 1500011D18Rik|AI315070|AIGP 2|KIAA1253|LRRGT00191|mKIAA1253|SERC1_HUMAN|SERINC1|Serine incorporator 1|TDE1L|TDE2|TMS 2|Tms2|Tumor differentially expressed 1 protein like|Tumor differentially expressed protein 1-like|Tumor differentially expressed protein 2|UNQ396/PRO732
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRX5
- Q9NRX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together