Anti-POLR2M Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: POLR2M
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77814-970
- 77814-969
- Antigen Symbol:
- POLR2M
- POLR2M
- Antigen Name:
- DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5 protein (2-91AA)
- DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5 protein (2-91AA)
- Antigen Synonyms:
- POLR2M|GRINL1A|DNA-directed RNA polymerase II subunit GRINL1A|isoforms 4/5|DNA-directed RNA polymerase II subunit M
- POLR2M|GRINL1A|DNA-directed RNA polymerase II subunit GRINL1A|isoforms 4/5|DNA-directed RNA polymerase II subunit M
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6EEV4
- Q6EEV4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together