Anti-MARK4 Rabbit Polyclonal Antibody
Catalog # 77807-267
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of MARK4
- Antigen Symbol:MARK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q96L34
- Antigen Synonyms:FLJ90097|KIAA1860|MAP/microtubule affinity regulating kinase 4|MAP/microtubule affinity regulating kinase 4L|MAP/microtubule affinity regulating kinase like 1|MAP/microtubule affinity-regulating kinase 4|MAP/microtubule affinity-regulating kinase-like 1|MARK 4|MARK 4L|MARK L1|MARK4|MARK4 serine/threonine protein kinase|MARK4_HUMAN|MARK4S|MARKL 1|MARKL1|Nbla00650|Putative protein product of Nbla00650
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:40000
Type:
Antigen: MARK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse