Order Entry
United States
ContactUsLinkComponent
Anti-HADHB Rabbit Recombinant Polyclonal Antibody
Anti-HADHB Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-HADHB Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77806-430
  • 77806-429
  • Antigen Symbol:
  • HADHB
  • HADHB
  • Antigen Name:
  • Trifunctional enzyme subunit beta, mitochondrial protein (35-283AA)
  • Trifunctional enzyme subunit beta, mitochondrial protein (35-283AA)
  • Antigen Synonyms:
  • 2 enoyl Coenzyme A (CoA) hydratase beta subunit|3 ketoacyl Coenzyme A (CoA) thiolase of mitochondrial trifunctional protein beta subunit|3 ketoacyl Coenzyme A thiolase|3 ketoacyl Coenzyme A thiolase of mitochondrial trifunctional protein beta subunit|3-ketoacyl-CoA thiolase|Acetyl CoA acyltransferase|Acetyl-CoA acyltransferase|Beta ketothiolase|Beta-ketothiolase|ECHB|ECHB_HUMAN|HADH|Hadhb|Hydroxyacyl CoA dehydrogenase/3 ketoacyl CoA thiolase/enoyl CoA hydratase (trifunctional protein)|beta subunit|Hydroxyacyl Coenzyme A (CoA) dehydrogenase beta subunit|Hydroxyacyl Coenzyme A dehydrogenase|Hydroxyacyl Coenzyme A dehydrogenase beta subunit|Hydroxyacyl Coenzyme A dehydrogenase/3 ketoacyl Coenzyme A thiolase/enoyl Coenzyme A hydratase (trifunctional protein) beta subunit|MGC87480|Mitochondrial trifunctional enzyme beta subunit|Mitochondrial trifunctional protein beta subunit|MSTP 029|MSTP029|MTPB|TP beta|TP-beta|TPbeta|Trifunctional enzyme subunit beta|Trifunctional enzyme subunit beta mitochondrial|Trifunctional protein
  • 2 enoyl Coenzyme A (CoA) hydratase beta subunit|3 ketoacyl Coenzyme A (CoA) thiolase of mitochondrial trifunctional protein beta subunit|3 ketoacyl Coenzyme A thiolase|3 ketoacyl Coenzyme A thiolase of mitochondrial trifunctional protein beta subunit|3-ketoacyl-CoA thiolase|Acetyl CoA acyltransferase|Acetyl-CoA acyltransferase|Beta ketothiolase|Beta-ketothiolase|ECHB|ECHB_HUMAN|HADH|Hadhb|Hydroxyacyl CoA dehydrogenase/3 ketoacyl CoA thiolase/enoyl CoA hydratase (trifunctional protein)|beta subunit|Hydroxyacyl Coenzyme A (CoA) dehydrogenase beta subunit|Hydroxyacyl Coenzyme A dehydrogenase|Hydroxyacyl Coenzyme A dehydrogenase beta subunit|Hydroxyacyl Coenzyme A dehydrogenase/3 ketoacyl Coenzyme A thiolase/enoyl Coenzyme A hydratase (trifunctional protein) beta subunit|MGC87480|Mitochondrial trifunctional enzyme beta subunit|Mitochondrial trifunctional protein beta subunit|MSTP 029|MSTP029|MTPB|TP beta|TP-beta|TPbeta|Trifunctional enzyme subunit beta|Trifunctional enzyme subunit beta mitochondrial|Trifunctional protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P55084
  • P55084
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together