Anti-TRIM62 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: TRIM62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-167
- 77820-166
- Antigen Symbol:
- TRIM62
- TRIM62
- Antigen Name:
- Synthetic peptide of TRIM62
- Synthetic peptide of TRIM62
- Antigen Synonyms:
- 6330414G21Rik|AI450348|DEAR1|ductal epithelium-associated RING Chromosome 1|E3 ubiquitin-protein ligase TRIM62|FLJ10759|FLJ16558|MGC115263|TRI62_HUMAN|TRIM62|Tripartite motif containing 62|Tripartite motif-containing protein 62
- 6330414G21Rik|AI450348|DEAR1|ductal epithelium-associated RING Chromosome 1|E3 ubiquitin-protein ligase TRIM62|FLJ10759|FLJ16558|MGC115263|TRI62_HUMAN|TRIM62|Tripartite motif containing 62|Tripartite motif-containing protein 62
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BVG3
- Q9BVG3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together