Anti-LY9 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LY9
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-170
- 77842-169
- Antigen Symbol:
- LY9
- LY9
- Antigen Name:
- T-lymphocyte surface antigen Ly-9 protein (160-360AA)
- T-lymphocyte surface antigen Ly-9 protein (160-360AA)
- Antigen Synonyms:
- CD229|Cell surface molecule Ly-9|Cell surface molecule Ly9|hly9|LY 9|Ly9|LY9_HUMAN|Lymphocyte antigen 9|mLY9|Signaling lymphocytic activation molecule 3|SLAM family member 3|SLAMF3|T lymphocyte surface antigen Ly9 precursor|T-lymphocyte surface antigen Ly-9
- CD229|Cell surface molecule Ly-9|Cell surface molecule Ly9|hly9|LY 9|Ly9|LY9_HUMAN|Lymphocyte antigen 9|mLY9|Signaling lymphocytic activation molecule 3|SLAM family member 3|SLAMF3|T lymphocyte surface antigen Ly9 precursor|T-lymphocyte surface antigen Ly-9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBG7
- Q9HBG7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together