Anti-MCCC1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: MCCC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-239
- 77833-240
- Antigen Symbol:
- MCCC1
- MCCC1
- Antigen Name:
- Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial protein (1-200AA)
- Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial protein (1-200AA)
- Antigen Synonyms:
- MCCC1|MCCAMethylcrotonoyl-CoA carboxylase subunit alpha|mitochondrial|MCCase subunit alpha|EC 6.4.1.4|3-methylcrotonyl-CoA carboxylase 1|3-methylcrotonyl-CoA carboxylase biotin-containing subunit|3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha
- MCCC1|MCCAMethylcrotonoyl-CoA carboxylase subunit alpha|mitochondrial|MCCase subunit alpha|EC 6.4.1.4|3-methylcrotonyl-CoA carboxylase 1|3-methylcrotonyl-CoA carboxylase biotin-containing subunit|3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RQ3
- Q96RQ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together