Anti-PPAT Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:500 to 1:2000, IHC: 1:100 to 1:300
Type:
Antigen: PPAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77814-416
- 77814-415
- Antigen Symbol:
- PPAT
- PPAT
- Antigen Name:
- Fusion protein of PPAT
- Fusion protein of PPAT
- Antigen Synonyms:
- Amidophosphoribosyltransferase [Precursor]|Amidophosphoribosyltransferase|ATase|Glutamine phosphoribosylpyrophosphatate amidotransferase|Glutamine phosphoribosylpyrophosphate amidotransferase|Glutamine PRPP amidotransferase|GPAT|Phosphoribosyl pyrophosphate amidotransferase|Ppat|PRAT|PUR1_HUMAN
- Amidophosphoribosyltransferase [Precursor]|Amidophosphoribosyltransferase|ATase|Glutamine phosphoribosylpyrophosphatate amidotransferase|Glutamine phosphoribosylpyrophosphate amidotransferase|Glutamine PRPP amidotransferase|GPAT|Phosphoribosyl pyrophosphate amidotransferase|Ppat|PRAT|PUR1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06203
- Q06203
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together