Order Entry
United States
ContactUsLinkComponent
Anti-PDE1C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-PDE1C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-PDE1C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77825-290
  • 77825-289
  • Antigen Symbol:
  • PDE1C
  • PDE1C
  • Antigen Name:
  • Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1C protein (542-634AA)
  • Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1C protein (542-634AA)
  • Antigen Synonyms:
  • 5''-cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin dependent 3'5' cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin-dependent 3''|Cam PDE 1C|Cam-PDE 1C|hCam 3|hCam-3|HCAM3|HSPDE1C1A|Human 3'5' cyclic nucleotide Phosphodiesterase|PDE 1C|Pde1c|PDE1C_HUMAN|Phosphodiesterase 1C|Phosphodiesterase 1C calmodulin dependent|Phosphodiesterase 1C calmodulin dependent 70kDa
  • 5''-cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin dependent 3'5' cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin-dependent 3''|Cam PDE 1C|Cam-PDE 1C|hCam 3|hCam-3|HCAM3|HSPDE1C1A|Human 3'5' cyclic nucleotide Phosphodiesterase|PDE 1C|Pde1c|PDE1C_HUMAN|Phosphodiesterase 1C|Phosphodiesterase 1C calmodulin dependent|Phosphodiesterase 1C calmodulin dependent 70kDa
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q14123
  • Q14123
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together