Anti-SKAP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SKAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-009
- 77832-010
- Antigen Symbol:
- SKAP1
- SKAP1
- Antigen Name:
- Src kinase-associated phosphoprotein 1 protein (1-358AA)
- Src kinase-associated phosphoprotein 1 protein (1-358AA)
- Antigen Synonyms:
- pp55|SCAP 1|SCAP1|SKAP 1|SKAP 55|SKAP-55|Skap1|SKAP1_HUMAN|SKAP55 adaptor protein|SRC family associated phosphoprotein 1|Src family-associated phosphoprotein 1|Src kinase associated phosphoprotein 1|SRC kinase associated phosphoprotein 55 kD|Src kinase associated phosphoprotein of 55 kDa|Src kinase-associated phosphoprotein 1|Src kinase-associated phosphoprotein of 55 kDa
- pp55|SCAP 1|SCAP1|SKAP 1|SKAP 55|SKAP-55|Skap1|SKAP1_HUMAN|SKAP55 adaptor protein|SRC family associated phosphoprotein 1|Src family-associated phosphoprotein 1|Src kinase associated phosphoprotein 1|SRC kinase associated phosphoprotein 55 kD|Src kinase associated phosphoprotein of 55 kDa|Src kinase-associated phosphoprotein 1|Src kinase-associated phosphoprotein of 55 kDa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WV1
- Q86WV1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together