Anti-GTF2I Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: GTF2I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77814-127
- 77814-128
- Antigen Symbol:
- GTF2I
- GTF2I
- Antigen Name:
- Synthetic peptide of GTF2I
- Synthetic peptide of GTF2I
- Antigen Synonyms:
- BAP 135|BAP-135|BAP135|Bruton tyrosine kinase associated protein 135|Bruton tyrosine kinase-associated protein 135|BTK associated protein 135|BTK associated protein 135kD|BTK associated protein|BTK-associated protein 135|BTKAP 1|BTKAP1|DIWS|FLJ38776|FLJ56355|General transcription factor II i|General transcription factor II-I|General transcription factor IIi|GTF 2I|Gtf2i|GTF2I_HUMAN|GTFII I|GTFII-I|IB 291|IB291|SPIN|SRF Phox 1 interacting protein|SRF Phox1 interacting protein|SRF-Phox1-interacting protein|TFII-I|Transcription factor II I|WBS|WBSCR 6|WBSCR6|Williams Beuren syndrome chromosome region 6|Williams Beuren syndrome chromosome region 6 protein|Williams-Beuren syndrome chromosomal region 6 protein
- BAP 135|BAP-135|BAP135|Bruton tyrosine kinase associated protein 135|Bruton tyrosine kinase-associated protein 135|BTK associated protein 135|BTK associated protein 135kD|BTK associated protein|BTK-associated protein 135|BTKAP 1|BTKAP1|DIWS|FLJ38776|FLJ56355|General transcription factor II i|General transcription factor II-I|General transcription factor IIi|GTF 2I|Gtf2i|GTF2I_HUMAN|GTFII I|GTFII-I|IB 291|IB291|SPIN|SRF Phox 1 interacting protein|SRF Phox1 interacting protein|SRF-Phox1-interacting protein|TFII-I|Transcription factor II I|WBS|WBSCR 6|WBSCR6|Williams Beuren syndrome chromosome region 6|Williams Beuren syndrome chromosome region 6 protein|Williams-Beuren syndrome chromosomal region 6 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78347
- P78347
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together