Anti-TMF1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TMF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77811-167
- 77803-167
- Antigen Symbol:
- TMF1
- TMF1
- Antigen Name:
- TATA element modulatory factor protein (878-986AA)
- TATA element modulatory factor protein (878-986AA)
- Antigen Synonyms:
- Androgen receptor coactivator 160 kDa protein|Androgen receptor-associated protein of 160 kDa|ARA160|TATA element modulatory factor 1|TATA element modulatory factor|TMF|Tmf1|TMF1_HUMAN
- Androgen receptor coactivator 160 kDa protein|Androgen receptor-associated protein of 160 kDa|ARA160|TATA element modulatory factor 1|TATA element modulatory factor|TMF|Tmf1|TMF1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P82094
- P82094
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together